Assay Details
| Gene Symbol: H1foo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138311.2 | NP_612184.1 | 2 - 3 | 397 | 100 | |
| GenBank mRNA | AK135944.1 | - | 2 - 3 | 401 | 100 | |
| AK162137.1 | - | 2 - 3 | 376 | 100 | ||
| AK162154.1 | - | 2 - 3 | 394 | 100 | ||
| AK165127.1 | - | 3 - 4 | 479 | 100 | ||
| AY007195.1 | - | 2 - 3 | 397 | 100 | ||
| BC137916.1 | - | 2 - 3 | 393 | 8860935 | 100 | |
| BC145046.1 | - | 2 - 3 | 393 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

H1foo
Genomic Map