Assay Details
| Gene Symbol: Coro1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011779.3 | NP_035909.2 | 7 - 8 | 963 | 93 | |
| GenBank mRNA | AF143957.1 | - | 7 - 8 | 944 | 93 | |
| AK077552.1 | - | 7 - 8 | 973 | 93 | ||
| AK079327.1 | - | 7 - 8 | 973 | 93 | ||
| AK082986.1 | - | 7 - 8 | 965 | 93 | ||
| AK088101.1 | - | 7 - 8 | 965 | 93 | ||
| AK151017.1 | - | 7 - 8 | 952 | 93 | ||
| AK151749.1 | - | 7 - 8 | 952 | 93 | ||
| AK159506.1 | - | 7 - 8 | 955 | 93 | ||
| AK167089.1 | - | 7 - 8 | 952 | 93 | ||
| AK169404.1 | - | 7 - 8 | 93 | |||
| AK191781.1 | - | 2 - 3 | 170 | 93 | ||
| AK197507.1 | - | 2 - 3 | 170 | 93 | ||
| BC009126.1 | - | 5 - 6 | 660 | 93 | ||
| BC019444.1 | - | 7 - 8 | 934 | 4167810 | 93 | |
| BC085498.1 | - | 7 - 8 | 933 | 5716526 | 93 | |
| BC087553.1 | - | 7 - 8 | 951 | 30460345 | 93 | |
| BC099671.1 | - | 7 - 8 | 899 | 4918161 | 93 | |
More Information
Set Membership: |
|

Coro1c
Genomic Map