Assay Details
| Gene Symbol: Apoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013474.2 | NP_038502.2 | 2 - 3 | 149 | 147 | |
| GenBank mRNA | AK145823.1 | - | 2 - 3 | 112 | 147 | |
| BC031786.1 | - | 1 - 2 | 95 | 4195958 | 147 | |
| BU609941.1 | - | 300 | 147 | |||
| BY780589.1 | - | 2 - 3 | 147 | 147 | ||
| CB947382.1 | - | 2 - 3 | 114 | 147 | ||
| S79794.1 | - | 1 - 2 | 55 | 147 | ||
| X04119.1 | - | 1 - 2 | 81 | 147 | ||
| X62772.1 | - | 2 - 3 | 97 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apoa2
Genomic Map