Assay Details
| Gene Symbol: Tcl1b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013776.1 | NP_038804.1 | 1 - 2 | 192 | 107 | |
| GenBank mRNA | AF195493.1 | - | 1 - 2 | 192 | 107 | |
| AK139627.1 | - | 1 - 2 | 207 | 107 | ||
| BC125294.1 | - | 1 - 2 | 189 | 40129809 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcl1b5
Genomic Map