Assay Details
| Gene Symbol: Tcl1b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013776.1 | NP_038804.1 | 3 - 4 | 450 | 92 | |
| GenBank mRNA | AF195493.1 | - | 3 - 4 | 450 | 92 | |
| AK139627.1 | - | 3 - 4 | 465 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tcl1b5
Genomic Map