Assay Details
| Gene Symbol: Bhmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016668.3 | NP_057877.1 | 4 - 5 | 737 | 104 | |
| GenBank mRNA | AF033381.1 | - | 4 - 5 | 480 | 104 | |
| AK077108.1 | - | 4 - 5 | 544 | 104 | ||
| AK145668.1 | - | 4 - 5 | 579 | 104 | ||
| AK149457.1 | - | 4 - 5 | 544 | 104 | ||
| AK166754.1 | - | 4 - 5 | 546 | 104 | ||
| AK166818.1 | - | 4 - 5 | 545 | 104 | ||
| BC037004.1 | - | 4 - 5 | 539 | 5100429 | 104 | |
| BC093510.1 | - | 4 - 5 | 512 | 5101184 | 104 | |
| BC110307.1 | - | 4 - 5 | 940 | 5310035 | 104 | |
| BG078929.2 | - | 2 - 3 | 229 | 104 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bhmt
Genomic Map