Assay Details
| Gene Symbol: Padi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153106.2 | NP_694746.2 | 11 - 12 | 1361 | 99 | |
| GenBank mRNA | AF529423.1 | - | 11 - 12 | 1337 | 99 | |
| AK139616.1 | - | 11 - 12 | 1362 | 99 | ||
| BC053724.1 | - | 11 - 12 | 1359 | 30051727 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Padi6
Genomic Map