| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100019623.mm.1 |
NM_153288.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019623.mm.1 |
NM_153288.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019623.mm.1 |
TC1100002037.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100019625.mm.1 |
NM_153288.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019625.mm.1 |
NM_153288.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019625.mm.1 |
TC1100002037.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100019617.mm.1 |
NM_153288.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019617.mm.1 |
NM_153288.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019617.mm.1 |
TC1100002037.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100019619.mm.1 |
NM_153288.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019619.mm.1 |
NM_153288.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019619.mm.1 |
TC1100002037.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100019624.mm.1 |
NM_153288.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019624.mm.1 |
NM_153288.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100019624.mm.1 |
TC1100002037.mm.1 |
MTA TC |