Assay Details
| Gene Symbol: U2af1l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170760.3 | NP_739566.1 | 2 - 3 | 149 | 72 | |
| GenBank mRNA | AF419339.1 | - | 2 - 3 | 133 | 72 | |
| BC051477.1 | - | 2 - 3 | 157 | 72 | ||
| BC060972.1 | - | 2 - 3 | 149 | 6593404 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

U2af1l4
Genomic Map