Assay Details
| Gene Symbol: U2af1l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170760.3 | NP_739566.1 | 4 - 5 | 257 | 86 | |
| GenBank mRNA | AF419339.1 | - | 4 - 5 | 241 | 86 | |
| BC051477.1 | - | 4 - 5 | 265 | 86 | ||
| BC060972.1 | - | 4 - 5 | 257 | 6593404 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

U2af1l4
Genomic Map