Assay Details
| Gene Symbol: Phyhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252568.2 | NP_001239497.1 | 1 - 2 | 424 | 93 | |
| NM_172267.4 | NP_758471.1 | 1 - 2 | 424 | 93 | ||
| GenBank mRNA | AK005293.1 | - | 1 - 2 | 425 | 93 | |
| AK138607.1 | - | 1 - 2 | 99 | 93 | ||
| BC132271.1 | - | 1 - 2 | 35 | 40130548 | 93 | |
| BC132273.1 | - | 1 - 2 | 35 | 40130550 | 93 | |
| BC145356.1 | - | 1 - 2 | 35 | 9053914 | 93 | |
| BC171925.1 | - | 1 - 2 | 35 | 93 | ||
| BY344206.1 | - | 1 - 2 | 78 | 93 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK005293.1,AK138607.1,BC132271.1,BC132273.1,BC145356.1,NM_001252568.2,NM_172267.4 transcripts.
Set Membership: |
|

Phyhd1
Genomic Map