Assay Details
| Gene Symbol: Phyhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252568.2 | NP_001239497.1 | 5 - 6 | 779 | 88 | |
| NM_001252570.1 | NP_001239499.1 | 4 - 5 | 400 | 88 | ||
| NM_001252571.1 | NP_001239500.1 | 4 - 5 | 400 | 88 | ||
| NM_001281829.1 | NP_001268758.1 | 5 - 6 | 426 | 88 | ||
| NM_172267.4 | NP_758471.1 | 5 - 6 | 779 | 88 | ||
| GenBank mRNA | AI182929.1 | - | 5 - 6 | 401 | 88 | |
| AK005293.1 | - | 5 - 6 | 780 | 88 | ||
| AK138607.1 | - | 5 - 6 | 454 | 88 | ||
| BC039982.1 | - | 5 - 6 | 421 | 88 | ||
| BC132271.1 | - | 5 - 6 | 390 | 40130548 | 88 | |
| BC132273.1 | - | 5 - 6 | 390 | 40130550 | 88 | |
| BC145356.1 | - | 5 - 6 | 390 | 9053914 | 88 | |
| BQ829083.1 | - | 4 - 5 | 396 | 88 | ||
| CX235839.1 | - | 3 - 4 | 246 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phyhd1
Genomic Map