Assay Details
| Gene Symbol: Phyhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252570.1 | NP_001239499.1 | 5 - 6 | 459 | 79 | |
| NM_001281829.1 | NP_001268758.1 | 6 - 7 | 485 | 79 | ||
| NM_172267.4 | NP_758471.1 | 6 - 7 | 838 | 79 | ||
| GenBank mRNA | AA620138.1 | - | 2 - 3 | 81 | 79 | |
| AK005293.1 | - | 6 - 7 | 839 | 79 | ||
| AK138607.1 | - | 6 - 7 | 513 | 79 | ||
| BC039982.1 | - | 6 - 7 | 480 | 79 | ||
| BC132271.1 | - | 6 - 7 | 449 | 40130548 | 79 | |
| BC132273.1 | - | 6 - 7 | 449 | 40130550 | 79 | |
| BQ829083.1 | - | 5 - 6 | 455 | 79 | ||
| CX235839.1 | - | 4 - 5 | 305 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phyhd1
Genomic Map