Assay Details
| Gene Symbol: Phyhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252570.1 | NP_001239499.1 | 7 - 8 | 545 | 93 | |
| NM_001281829.1 | NP_001268758.1 | 8 - 9 | 571 | 93 | ||
| NM_172267.4 | NP_758471.1 | 8 - 9 | 924 | 93 | ||
| GenBank mRNA | AK005293.1 | - | 8 - 9 | 925 | 93 | |
| AK138607.1 | - | 8 - 9 | 599 | 93 | ||
| BC039982.1 | - | 8 - 9 | 566 | 93 | ||
| BC132271.1 | - | 8 - 9 | 535 | 40130548 | 93 | |
| BC132273.1 | - | 8 - 9 | 535 | 40130550 | 93 | |
| CX235839.1 | - | 6 - 7 | 391 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phyhd1
Genomic Map