Assay Details
| Gene Symbol: Phyhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252568.2 | NP_001239497.1 | 8 - 9 | 989 | 103 | |
| NM_001252570.1 | NP_001239499.1 | 8 - 9 | 679 | 103 | ||
| NM_001252571.1 | NP_001239500.1 | 7 - 8 | 616 | 103 | ||
| NM_001281829.1 | NP_001268758.1 | 9 - 10 | 705 | 103 | ||
| NM_172267.4 | NP_758471.1 | 9 - 10 | 1058 | 103 | ||
| GenBank mRNA | AA620138.1 | - | 5 - 6 | 300 | 103 | |
| AK005293.1 | - | 9 - 10 | 1059 | 103 | ||
| AK138607.1 | - | 9 - 10 | 733 | 103 | ||
| BC039982.1 | - | 9 - 10 | 700 | 103 | ||
| BC132271.1 | - | 9 - 10 | 669 | 40130548 | 103 | |
| BC132273.1 | - | 9 - 10 | 669 | 40130550 | 103 | |
| BC145356.1 | - | 8 - 9 | 600 | 9053914 | 103 | |
| BC171925.1 | - | 4 - 5 | 404 | 103 | ||
| CO041185.1 | - | 3 - 4 | 103 | |||
| CX235839.1 | - | 7 - 8 | 525 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phyhd1
Genomic Map