Assay Details
| Gene Symbol: Pced1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172293.4 | NP_758497.1 | 2 - 3 | 140 | 98 | |
| GenBank mRNA | AK051480.1 | - | 2 - 3 | 141 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK051480.1, NM_172293.4 transcripts.
Set Membership: |
|

Pced1b
Genomic Map