Assay Details
| Gene Symbol: Gpkow | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173747.3 | NP_776108.2 | 10 - 11 | 1425 | 88 | |
| GenBank mRNA | AK028433.1 | - | 10 - 11 | 1347 | 88 | |
| AK144167.1 | - | 10 - 11 | 1356 | 88 | ||
| BC062646.1 | - | 8 - 9 | 975 | 88 | ||
| BC092224.1 | - | 10 - 11 | 1331 | 30355916 | 88 | |
| BC120679.1 | - | 9 - 10 | 1108 | 40129602 | 88 | |
| BC125335.1 | - | 9 - 10 | 1108 | 40129850 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239704 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239703 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239707 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239706 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045602 | BI076639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045602 | BC062646.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045602 | BC092224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045602 | AK028433.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045602 | AK144167.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610904 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814537 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105152 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169088 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157326 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045602 | BI076639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045602 | BC062646.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045602 | BC092224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045602 | AK028433.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045602 | AK144167.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32239720 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09226557 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169088 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157326 | - | - |
| Whole Mouse Genome G4122A | A_51_P314584 | BI076639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314584 | BC062646.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314584 | BC092224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314584 | AK028433.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P314584 | AK144167.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P254884 | AK017864.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120279 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157326 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169088 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15089789 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15089792 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239703 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239704 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239707 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32239706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32239761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14990724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15011795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15029901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14678583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14673930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14827711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14707230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14720718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15107197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14723936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14827715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32239687 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10598383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10598382 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434344_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458023_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10598381 | - | - |
| Mouse Genome 430 2.0 Array | 1434344_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458023_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458023_at | - | - |
| Mouse Genome 430 2.0 Array | 1434344_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 209416_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 209416_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00000352.mm.1 | NM_173747.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000352.mm.1 | NM_173747.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000352.mm.1 | TC0X00000063.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000353.mm.1 | NM_173747.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000353.mm.1 | NM_173747.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000353.mm.1 | TC0X00000063.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00000355.mm.1 | NM_173747.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000355.mm.1 | NM_173747.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00000355.mm.1 | TC0X00000063.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434344_at | - | - |
| Mouse Expression Set 430 | 1434344_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458023_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434344_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458023_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434344_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6940341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6940341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3800138 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006940341 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800138 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245453 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2728189 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2728189 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245453 | - | - |
| mouse-6 v1.1 (Illumina) | scl00209416.1_66-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0209416.11_89-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800138 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6940341 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00209416.1_66-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0209416.11_89-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800138 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940341 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2728189 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245453 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245453 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2728189 | - | - |

Gpkow
Genomic Map