Assay Details
| Gene Symbol: Gpkow | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173747.3 | NP_776108.2 | 6 - 7 | 1005 | 85 | |
| GenBank mRNA | AK028433.1 | - | 6 - 7 | 927 | 85 | |
| AK144167.1 | - | 6 - 7 | 935 | 85 | ||
| BC062646.1 | - | 4 - 5 | 555 | 85 | ||
| BC092224.1 | - | 6 - 7 | 911 | 30355916 | 85 | |
| BC120679.1 | - | 5 - 6 | 688 | 40129602 | 85 | |
| BC125335.1 | - | 5 - 6 | 688 | 40129850 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpkow
Genomic Map