Assay Details
| Gene Symbol: Gpkow | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173747.3 | NP_776108.2 | 8 - 9 | 1269 | 100 | |
| GenBank mRNA | AK028433.1 | - | 8 - 9 | 1191 | 100 | |
| AK144167.1 | - | 8 - 9 | 1200 | 100 | ||
| BC062646.1 | - | 6 - 7 | 819 | 100 | ||
| BC092224.1 | - | 8 - 9 | 1175 | 30355916 | 100 | |
| BC120679.1 | - | 7 - 8 | 952 | 40129602 | 100 | |
| BC125335.1 | - | 7 - 8 | 952 | 40129850 | 100 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpkow
Genomic Map