Assay Details
| Gene Symbol: Gpkow | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173747.3 | NP_776108.2 | 9 - 10 | 1334 | 96 | |
| GenBank mRNA | AK028433.1 | - | 9 - 10 | 1256 | 96 | |
| AK144167.1 | - | 9 - 10 | 1265 | 96 | ||
| BC062646.1 | - | 7 - 8 | 884 | 96 | ||
| BC092224.1 | - | 9 - 10 | 1240 | 30355916 | 96 | |
| BC120679.1 | - | 8 - 9 | 1017 | 40129602 | 96 | |
| BC125335.1 | - | 8 - 9 | 1017 | 40129850 | 96 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpkow
Genomic Map