Assay Details
| Gene Symbol: Tmem139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175408.4 | NP_780617.2 | 1 - 2 | 55 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_175408.4 transcripts.
Set Membership: |
|

Tmem139
Genomic Map