| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00007135.mm.1 |
NM_175442.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007135.mm.1 |
NM_175442.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007135.mm.1 |
TC0X00001084.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00007138.mm.1 |
NM_001122595.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007138.mm.1 |
NM_001122595.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007138.mm.1 |
NM_001122596.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007138.mm.1 |
NM_001122596.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007138.mm.1 |
NM_175442.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007138.mm.1 |
NM_175442.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007138.mm.1 |
TC0X00001084.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00007137.mm.1 |
NM_001122595.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007137.mm.1 |
NM_001122595.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007137.mm.1 |
NM_001122596.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007137.mm.1 |
NM_001122596.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007137.mm.1 |
NM_175442.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007137.mm.1 |
NM_175442.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007137.mm.1 |
TC0X00001084.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00007139.mm.1 |
NM_001122595.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007139.mm.1 |
NM_001122595.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007139.mm.1 |
NM_001122596.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007139.mm.1 |
NM_001122596.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007139.mm.1 |
NM_175442.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007139.mm.1 |
NM_175442.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00007139.mm.1 |
TC0X00001084.mm.1 |
MTA TC |