Assay Details
| Gene Symbol: Hist1h2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175664.3 | NP_783595.1 | 1 | 45 | 70 | |
| GenBank mRNA | BC116853.1 | - | 1 | 45 | 40126172 | 70 |
| BC116857.1 | - | 1 | 45 | 40126176 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC116853.1, BC116857.1, NM_175664.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hist1h2bb
Genomic Map