Assay Details
| Gene Symbol: Dnajc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176835.2 | NP_789805.1 | 2 - 3 | 1071 | 110 | |
| GenBank mRNA | AK013323.1 | - | 2 - 3 | 1094 | 110 | |
| AK050294.1 | - | 2 - 3 | 1050 | 110 | ||
| BC039633.1 | - | 2 - 3 | 1071 | 5136672 | 110 | |
More Information
Set Membership: |
|

Dnajc22
Genomic Map