Assay Details
| Gene Symbol: Slc35e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177186.4 | NP_796160.1 | 5 - 6 | 1195 | 113 | |
| GenBank mRNA | AK048716.1 | - | 5 - 6 | 1064 | 113 | |
| AK145065.1 | - | 6 - 7 | 1118 | 113 | ||
| AK157354.1 | - | 5 - 6 | 1091 | 113 | ||
| BC058728.1 | - | 5 - 6 | 1195 | 6407259 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35e2
Genomic Map