Assay Details
| Gene Symbol: Slc35e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177186.4 | NP_796160.1 | 7 - 8 | 1308 | 103 | |
| GenBank mRNA | AK048716.1 | - | 7 - 8 | 1177 | 103 | |
| AK145065.1 | - | 8 - 9 | 103 | |||
| AK157354.1 | - | 7 - 8 | 1204 | 103 | ||
| BC058728.1 | - | 7 - 8 | 1308 | 6407259 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35e2
Genomic Map