Assay Details
| Gene Symbol: Ldoc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177630.3 | NP_808298.2 | 1 | 1147 | 95 | |
| GenBank mRNA | AK083881.1 | - | 1 | 1109 | 95 | |
| BC035491.1 | - | 1 | 203 | 95 | ||
| BC058638.1 | - | 1 | 816 | 6418256 | 95 | |
| BC094559.1 | - | 1 | 1147 | 6826763 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ldoc1l
Genomic Map