Assay Details
| Gene Symbol: Hic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178922.3 | NP_849253.2 | 1 - 2 | 148 | 86 | |
| GenBank mRNA | AK084975.1 | - | 1 - 2 | 71 | 86 | |
| AK129266.1 | - | 1 - 2 | 134 | 86 | ||
| AK156332.1 | - | 1 - 2 | 164 | 86 | ||
| AK157276.1 | - | 1 - 2 | 164 | 86 | ||
| BC065124.1 | - | 147 | 30537019 | 86 | ||
| BQ965108.1 | - | 1 - 2 | 155 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK084975.1, AK156332.1, AK157276.1, BC065124.1, NM_178922.3 transcripts.
Set Membership: |
|

Hic2
Genomic Map