| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600001713.mm.1 |
NM_178922.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001713.mm.1 |
NM_178922.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001713.mm.1 |
TC1600000212.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600001714.mm.1 |
NM_178922.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001714.mm.1 |
NM_178922.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001714.mm.1 |
TC1600000212.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600001715.mm.1 |
NM_178922.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001715.mm.1 |
NM_178922.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001715.mm.1 |
TC1600000212.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600001712.mm.1 |
NM_178922.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001712.mm.1 |
NM_178922.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001712.mm.1 |
TC1600000212.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600001716.mm.1 |
NM_178922.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001716.mm.1 |
NM_178922.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600001716.mm.1 |
TC1600000212.mm.1 |
MTA TC |