Assay Details
| Gene Symbol: Eras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181548.2 | NP_853526.1 | 1 - 2 | 143 | 102 | |
| GenBank mRNA | AB093573.1 | - | 1 - 2 | 143 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB093573.1, NM_181548.2 transcripts.
Set Membership: |
|

Eras
Genomic Map