Assay Details
| Gene Symbol: Nanos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194064.2 | NP_918953.2 | 1 | 448 | 82 | |
| GenBank mRNA | AB095972.1 | - | 1 | 448 | 82 | |
| JQ936992.1 | - | 1 | 378 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nanos2
Genomic Map