| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800023106.mm.1 |
NM_026572.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023106.mm.1 |
NM_026572.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023106.mm.1 |
TC0800003008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023104.mm.1 |
NM_026572.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023104.mm.1 |
NM_026572.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023104.mm.1 |
TC0800003008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023105.mm.1 |
NM_026572.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023105.mm.1 |
NM_026572.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023105.mm.1 |
TC0800003008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023107.mm.1 |
NM_026572.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023107.mm.1 |
NM_026572.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023107.mm.1 |
TC0800003008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023108.mm.1 |
NM_026572.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023108.mm.1 |
NM_026572.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023108.mm.1 |
TC0800003008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023111.mm.1 |
NM_026572.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023111.mm.1 |
NM_026572.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023111.mm.1 |
TC0800003008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800023109.mm.1 |
NM_026572.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023109.mm.1 |
NM_026572.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800023109.mm.1 |
TC0800003008.mm.1 |
MTA TC |