Assay Details
| Gene Symbol: Tra2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009186.4 | NP_033212.1 | 1 | 51 | 102 | |
| GenBank mRNA | AK049573.1 | - | 1 | 52 | 102 | |
| AK077418.1 | - | 1 | 49 | 102 | ||
| AK080378.1 | - | 1 | 41 | 102 | ||
| AK150007.1 | - | 1 | 52 | 102 | ||
| AK152519.1 | - | 1 | 46 | 102 | ||
| AK152761.1 | - | 1 | 52 | 102 | ||
| AK167387.1 | - | 1 | 50 | 102 | ||
| AK167544.1 | - | 1 | 52 | 102 | ||
| AK169483.1 | - | 1 | 41 | 102 | ||
| BC061177.1 | - | 1 | 48 | 6594226 | 102 | |
| BC086795.1 | - | 1 | 38 | 6828358 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049573.1, AK077418.1, AK080378.1, BC061177.1, BC086795.1, NM_009186.4 transcripts.
Set Membership: |
|

Tra2b
Genomic Map