Assay Details
| Gene Symbol: Gdf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013527.1 | NP_038555.1 | 1 | 48 | 93 | |
| GenBank mRNA | AF525752.1 | - | 1 | 48 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF525752.1, NM_013527.1 transcripts.
Set Membership: |
|

Gdf7
Genomic Map