| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00002952.mm.1 |
NM_173376.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002952.mm.1 |
NM_173376.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002952.mm.1 |
TC0X00000439.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002956.mm.1 |
NM_173376.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002956.mm.1 |
NM_173376.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002956.mm.1 |
TC0X00000439.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002953.mm.1 |
NM_173376.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002953.mm.1 |
NM_173376.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002953.mm.1 |
TC0X00000439.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002957.mm.1 |
NM_173376.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002957.mm.1 |
NM_173376.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002957.mm.1 |
TC0X00000439.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002954.mm.1 |
NM_173376.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002954.mm.1 |
NM_173376.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002954.mm.1 |
TC0X00000439.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002951.mm.1 |
NM_173376.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002951.mm.1 |
NM_173376.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002951.mm.1 |
TC0X00000439.mm.1 |
MTA TC |