Assay Details
| Gene Symbol: Apol9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173743.4 | NP_776104.1 | 1 | 48 | 82 | |
| GenBank mRNA | AK050167.1 | - | 1 | 50 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050167.1,NM_173743.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Apol9b
Genomic Map