Assay Details
| Gene Symbol: Mtch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019758.2 | NP_062732.1 | 1 | 147 | 94 | |
| GenBank mRNA | AK008712.1 | - | 1 | 148 | 94 | |
| AK011814.1 | - | 1 | 72 | 94 | ||
| AK016000.1 | - | 1 | 112 | 94 | ||
| BC028929.1 | - | 1 | 124 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008712.1, AK011814.1, NM_019758.2 transcripts.
Set Membership: |
|

Mtch2
Genomic Map