Assay Details
| Gene Symbol: Klrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021378.1 | NP_067353.1 | 2 | 254 | 85 | |
| GenBank mRNA | AF195779.1 | - | 2 | 254 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klrc3
Genomic Map