Assay Details
| Gene Symbol: ATP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | NC_005089_ATP8.0 | - | 42 | 108 | ||
More Information
Additional Information:
NC_005089 (strand:+; 7766..7969); ATP synthase F0 subunit 8
Set Membership: |
|

ATP8
Genomic Map