Assay Details
| Gene Symbol: Mir27b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029531.1 | 1 | 37 | 69 | ||
| Gene Symbol: 2010111I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289926.1 | NP_001276855.1 | 16 | 4636 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 23b | Mir23b |
Mirn23b, mmu-mir-23b |
-155.0 | Sense | Chr. 13 - 63300484 - 63300557 on NCBI Build GRCm38 |
| Downstream |
Fanconi anemia, complementation group C |
Fancc |
Facc, mir-3074-1 |
350.0 | Anti-sense | Chr. 13 - 63301134 - 63497290 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir27b
Genomic Map