Assay Details
| Gene Symbol: 9430060I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015525.1 | 1 - 2 | 114 | 81 | ||
| GenBank mRNA | BC090260.1 | - | 1 - 2 | 114 | 81 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BQ922611.1 | - | 113 | Chr. - - [-] on Build | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calpain 10 | Capn10 |
AW049679, Capn8, mKIAA1845 |
Anti-sense | Chr. 1 - 92934408 - 92947948 on NCBI Build GRCm38 | |
| Downstream | G protein-coupled receptor 35 | Gpr35 | KPG_007 | 272.0 | Anti-sense | Chr. 1 - 92950862 - 92986391 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

9430060I03Rik
Genomic Map