Assay Details
| Gene Symbol: Gm2518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015538.1 | 1 | 2254 | 101 | ||
| GenBank mRNA | AK082962.1 | - | 1 | 2253 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor |
Taf6l |
2810417N14Rik, BB223262, C530024J06Rik, Paf65a |
Anti-sense | Chr. 19 - 8774354 - 8786417 on NCBI Build GRCm38 | |
| Downstream | polymerase (RNA) II (DNA directed) polypeptide G | Polr2g |
2410046K11Rik, A230108L04Rik, C76415, RBP7, Rpo2-7l |
16.2kb | Anti-sense | Chr. 19 - 8793129 - 8798557 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm2518
Genomic Map