Assay Details
| Gene Symbol: A930007I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015567.2 | 2 - 3 | 219 | 57 | ||
| GenBank mRNA | AK137192.1 | - | 2 - 3 | 221 | 57 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | programmed cell death 1 ligand 2 | Pdcd1lg2 |
B7-DC, Btdc, F730015O22Rik, PD-L2 |
-37.2kb | Anti-sense | Chr. 19 - 29406240 - 29472927 on NCBI Build GRCm38 |
| Downstream | RAB6A GEF complex partner 1 | Ric1 |
C030046E11Rik, C130057E09Rik, Kiaa1432 |
1.3kb | Anti-sense | Chr. 19 - 29522282 - 29605921 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A930007I19Rik
Genomic Map