Assay Details
| Gene Symbol: Gm20268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037989.1 | 2 - 3 | 321 | 87 | ||
| GenBank mRNA | AK076805.1 | - | 2 - 3 | 283 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | contactin associated protein-like 5B | Cntnap5b |
C230078M14Rik, Caspr5-2 |
-682.6kb | Sense | Chr. 1 - 99772247 - 100486066 on NCBI Build GRCm38 |
| Downstream | cadherin 20 | Cdh20 | Cdh7 | 3.6mb | Sense | Chr. 1 - 104768819 - 104995481 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm20268
Genomic Map