Assay Details
Gene Symbol: Gm10638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027829.1 | 1 | 1243 | 85 | ||
GenBank mRNA | AI594960.1 | - | 1 | 85 | ||
AK133595.1 | - | 769 | 85 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AA060875.1 | - | 1 | Chr. 8 - 86746571 - 86747060 [-] on Build GRCm38 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | seven in absentia 1A | Siah1a |
AA982064, AI853500, Sinh1a |
Anti-sense | Chr. 8 - 86723920 - 86746006 on NCBI Build GRCm38 | |
Downstream | NEDD4 binding protein 1 | N4bp1 |
AI481586, C81621 |
94.1kb | Anti-sense | Chr. 8 - 86841136 - 86885258 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
