Assay Details
| Gene Symbol: 1110020A21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027930.1 | 1 - 2 | 514 | 109 | ||
| GenBank mRNA | BI737205.1 | - | 1 - 2 | 308 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein phosphatase 1B, magnesium dependent, beta isoform |
Ppm1b | PP2CB | Anti-sense | Chr. 17 - 84956741 - 85023992 on NCBI Build GRCm38 | |
| Downstream |
solute carrier family 3, member 1 |
Slc3a1 |
D2H, NBAT, NTAA, RBAT |
70.6kb | Anti-sense | Chr. 17 - 85028347 - 85064241 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107745 | NR_027930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107745 | AK003841.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107745 | NR_027929.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107745 | BI737205.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513940 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770086 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311042 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812319 | AK044317.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723948 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741006 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 698134 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676116 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227720 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489076 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 154761 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08875171 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03385814 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08875260 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03385841 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03385814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13325524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13077641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13213283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13072167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13288495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13383991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13410014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13575288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13362716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13120060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13264431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31407552 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1439574_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439574_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439574_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68531_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68531_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700024154.mm.1 | NR_027929.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024154.mm.1 | NR_027929.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024154.mm.1 | NR_027930.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024154.mm.1 | NR_027930.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024154.mm.1 | TC1700002658.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700024151.mm.1 | NR_027930.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024151.mm.1 | NR_027930.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024151.mm.1 | TC1700002658.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700024153.mm.1 | NR_027929.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024153.mm.1 | NR_027929.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024153.mm.1 | NR_027930.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024153.mm.1 | NR_027930.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024153.mm.1 | TC1700002658.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700024155.mm.1 | NR_027930.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024155.mm.1 | NR_027930.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700024155.mm.1 | TC1700002658.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439574_at | - | - |
| Mouse Expression Set 430 | 1439574_PM_at | - | - |
| Murine Genome U74v2 Set | 171481_at | - | - |
| Murine Genome U74v2 Set | 171481_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439574_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439574_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6660066 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2663960 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2663960 | - | - |

1110020A21Rik
Genomic Map