Assay Details
| Gene Symbol: Snord89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028555.2 | 1 | 49 | 65 | ||
| GenBank mRNA | AF357353.1 | - | 1 | 48 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
CCR4-NOT transcription complex, subunit 11 |
Cnot11 |
2410015L18Rik, C40, D1Bwg0212e |
-1.9kb | Anti-sense | Chr. 1 - 39535802 - 39546881 on NCBI Build GRCm38 |
| Downstream | ring finger protein 149 | Rnf149 |
1600023E10Rik, AV037261, C76763, Gm15832, Greul4, OTTMUSG00000026591 |
2.5kb | Sense | Chr. 1 - 39551294 - 39577441 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snord89
Genomic Map