Assay Details
| Gene Symbol: Snora16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029412.1 | 1 | 69 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small nucleolar RNA host gene 12 | Snhg12 | 2310005L22Rik | Sense | Chr. 4 - 132308678 - 132311024 on NCBI Build GRCm38 | |
| Downstream |
small nucleolar RNA, H/ACA box 44 |
Snora44 | MBI-64 | 353.0 | Sense | Chr. 4 - 132309953 - 132310069 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snora16a
Genomic Map