Assay Details
| Gene Symbol: 4933407K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029443.1 | 1 - 2 | 172 | 75 | ||
| GenBank mRNA | AK030939.1 | - | 1 - 2 | 173 | 75 | |
| AK053637.1 | - | 1 - 2 | 123 | 75 | ||
| AK163097.1 | - | 172 | 75 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB39B, member RAS oncogene family |
Rab39b | 6330580M05Rik | -182.0kb | Sense | Chr. X - 75572045 - 75578231 on NCBI Build GRCm38 |
| Downstream | plastin 3 (T-isoform) | Pls3 |
AI115446, AL024105, T-fimbrin |
21.0kb | Sense | Chr. X - 75785654 - 75875170 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933407K13Rik
Genomic Map