Assay Details
| Gene Symbol: 4933407K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029443.1 | 3 - 4 | 494 | 65 | ||
| GenBank mRNA | AK016728.1 | - | 1 - 2 | 230 | 65 | |
| AK030939.1 | - | 3 - 4 | 495 | 65 | ||
| AK053637.1 | - | 3 - 4 | 445 | 65 | ||
| AK163097.1 | - | 494 | 65 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB39B, member RAS oncogene family |
Rab39b | 6330580M05Rik | -173.3kb | Sense | Chr. X - 75572045 - 75578231 on NCBI Build GRCm38 |
| Downstream | plastin 3 (T-isoform) | Pls3 |
AI115446, AL024105, T-fimbrin |
30.1kb | Sense | Chr. X - 75785654 - 75875170 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933407K13Rik
Genomic Map